missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12442-2-AP-20UL
This item is not returnable.
View return policy
Description
Cyclic nucleotide phosphodiesterases play a role in signal transduction by regulating intracellular cyclic nucleotide concentrations through hydrolysis of cAMP and/or cGMP to their respective nucleoside 5-prime monophosphates. Members of the PDE1 family, such as PDE1A, are Cathat are activated by calmodulin in the presence of Ca.Specifications
PDE1A | |
Polyclonal | |
Unconjugated | |
PDE1A | |
61 kDa Cam PDE, Cam PDE 1A, hCam 1, HCAM1, HSPDE1A, PDE1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18573, 5136 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P54750, Q61481 | |
PDE1A | |
PDE1A Fusion Protein Ag3157 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PDE1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction