Learn More
PDE1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12442-2-AP-150UL
 View more versions of this product
Product Code. 16880154
  • 4652.90 SEK / 150µL
Description
Cyclic nucleotide phosphodiesterases play a role in signal transduction by regulating intracellular cyclic nucleotide concentrations through hydrolysis of cAMP and/or cGMP to their respective nucleoside 5-prime monophosphates. Members of the PDE1 family, such as PDE1A, are Cathat are activated by calmodulin in the presence of Ca.
Specifications
Specifications
PDE1A
Polyclonal
Unconjugated
PDE1A
61 kDa Cam PDE, Cam PDE 1A, hCam 1, HCAM1, HSPDE1A, PDE1A
Rabbit
Antigen Affinity Chromatography
RUO
18573, 5136
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P54750, Q61481
PDE1A
PDE1A Fusion Protein Ag3157
150 μL
Primary
Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PDE1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated