Learn More
PDE9A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12648-1-AP-150UL
 View more versions of this product
Product Code. 16880474
  • 4652.90 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The protein encoded by this gene catalyzes the hydrolysis of cAMP and cGMP to their corresponding monophosphates. The encoded protein plays a role in signal transduction by regulating the intracellular concentration of these cyclic nucleotides. Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifications
Specifications
PDE9A
Polyclonal
Unconjugated
PDE9A
HSPDE9A2, PDE9A, phosphodiesterase 9A
Rabbit
Antigen Affinity Chromatography
RUO
18585, 191569, 5152
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70628, O76083, Q8QZV1
PDE9A
PDE9A Fusion Protein Ag3372
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PDE9A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated