missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDIA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18233-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PDIA6.
P5 is a novel protein that was originally identified in humans. Functionally, P5 has been shown to have peptide binding ability and chaperone activity specific to certain proteins. P5 shares several structural similarities to PDI, an important protein of the ER. The deduced amino acid sequence of P5 shows that it contains the ER retention signal KDEL. P5 also contains two CXXC-like motifs, a motif responsible for oxidoreductase activity. These two CXXC motifs are found on the N- and C-terminus of the protein, one at each end. Studies suggest that the N-terminal CXXC motif is more important than the second for isomerase activity. Contrary to most of the PDI-like proteins, P5 is not stress-inducible.Specifications
PDIA6 | |
Polyclonal | |
Unconjugated | |
PDIA6 | |
ER protein 5, ERP5, P5, PDIA6, Protein disulfide isomerase A6, Protein disulfide isomerase P5, TXNDC7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10130, 286906, 71853 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15084, Q63081, Q922R8 | |
Pdia6 | |
PDIA6 Fusion Protein Ag12940 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |