Learn More
PDK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15647-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16884904
  • 3665.00 SEK / 150µL
Estimated Shipment: 31-03-2023
to see stock.
Description
Description
PDK2 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
Specifications
Specifications
PDK2
Polyclonal
Unconjugated
PDK2
PDHK2, PDK2
Rabbit
Antigen Affinity Chromatography
RUO
18604, 5164, 81530
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15119, Q64536, Q9JK42
PDK2
PDK2 Fusion Protein Ag8183
150 μL
Primary
Rat, Mouse, Human