Learn More
PDLIM5 Mouse anti-Human, Unconjugated, Clone: 1H9C9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67651-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16880396
  • 1355.00 SEK / 20µL
Estimated Shipment: 27-03-2023
to see stock.
Description
Description
The PDLIM5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PDLIM5. This antibody recognizes human antigen. The PDLIM5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PDLIM5
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot
1H9C9
Unconjugated
20 μL
10611
IgM