Learn More
PDLIM5 Mouse anti-Human, Unconjugated, Clone: 1H9C9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67651-1-IG-20UL
 View more versions of this product
Product Code. 16880396
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The PDLIM5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PDLIM5. This antibody recognizes human antigen. The PDLIM5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PDLIM5
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot
1H9C9
Unconjugated
20 μL
10611
IgM
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PDLIM5 Mouse anti-Human, Unconjugated, Clone: 1H9C9, Proteintech > 20 μL