missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PECR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14901-1-AP-20UL
This item is not returnable.
View return policy
Description
PECR (Peroxisomal Trans-2-Enoyl-CoA Reductase) is a Protein Coding gene. Among its related pathways are Metabolism and Allograft rejection. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and coenzyme binding. An important paralog of this gene is DECR1.Specifications
PECR | |
Polyclonal | |
Unconjugated | |
PECR | |
DCR RP, HPDHase, HSA250303, PECR, pVI ARL, SDR29C1, TERP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
111175, 113956, 55825 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99MZ7, Q9BY49, Q9WVK3 | |
PECR | |
PECR Fusion Protein Ag6693 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PECR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction