missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEDF Mouse anti-Human, Porcine, Clone: 2C5C8, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PEDF |
---|---|
Clone | 2C5C8 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888535
|
Proteintech
66554-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878535
|
Proteintech
66554-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PEDF gene is a member of the serpin gene family. Serpins are a group of serine protease inhibitors, some of which have also been reported to exhibit neurotrophic activity.Specifications
PEDF | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EPC 1, PEDF, PIG35, Serpin F1, SERPINF1 | |
SERPINF1 | |
IgG1 | |
Protein G | |
Antibody |
2C5C8 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
P36955 | |
5176, 780402 | |
PEDF Fusion Protein Ag26660 | |
Primary | |
-20°C | |
SERPINF1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PEDF Mouse anti-Human, Porcine, Clone: 2C5C8, Proteintech