Learn More
Hamilton™ PEEK HPLC Guard Column Starter Kit
Brand: Hamilton™ 79831
This item is not returnable.
View return policy
Specifications
7 μm | |
HPLC Guard Column Starter Kit | |
PEEK |
Product Content Correction
Product Title
Hamilton™ PEEK HPLC Guard Column Starter Kit > Particle Size: 7μm
Spot an opportunity for improvement?Share a Content Correction