Learn More
PEF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10151-1-AP-20UL
 View more versions of this product
Product Code. 16886343
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2. PEF1 is a Ca(2+)-binding protein that belongs to the penta-EF-hand (PEF) protein family, which includes the calpain small subunit (CAPNS1), sorcin (SRI), grancalcin (GCA), and ALG2 (PDCD6) (Kitaura et al., 2001 ).
Specifications
Specifications
PEF1
Polyclonal
Unconjugated
PEF1
ABP32, PEF1, PEF1A, PEFLIN
Rabbit
Antigen Affinity Chromatography
RUO
553115
-20°C
Liquid
Western Blot, Immunoprecipitation
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UBV8
PEF1
PEF1 Fusion Protein Ag0198
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PEF1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated