missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16097-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Scaffold protein which probably links Toll-like receptors (TLRs) to basic cellular processes via its interaction with the complex containing IRAK kinases and TRAF6. Can activate the MAP (mitogen activated protein) kinase pathway leading to activation of ELK1. Not required for NF-kappa-B activation.Specifications
PELI2 | |
Polyclonal | |
Unconjugated | |
PELI2 | |
E3 ubiquitin-protein ligase pellino homolog 2, PELI2, Pellino 2, pellino homolog 2 (Drosophila), Protein pellino homolog 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305835, 57161, 93834 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BST6, Q9HAT8 | |
PELI2 | |
PELI2 Fusion Protein Ag8940 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |