Learn More
Fisherbrand™ Penetration pH Electrodes
Useful for measuring potential of hydrogen ions. Fisherbrand™ Penetration pH Electrodes can be placed deep within samples.
Brand: Fisherbrand™
Additional Details : Weight : 0.10000kg
Description
- Reinforced
- Excellent for meat, cheese, fish, fruit
- Electrolyte KCI 3M + AgCl
- pH range of 0 to 12
- Temperature range of 0° to 80°C
Specifications
BNC | |
0°C to 80°C | |
25 mm |
0 to 12 | |
Standard | |
5 mm |