Learn More
Fisherbrand™ Penetration pH Electrodes
Useful for measuring potential of hydrogen ions. Fisherbrand™ Penetration pH Electrodes can be placed deep within samples.
2555.00 SEK
Specifications
pH Range | 0 to 12 |
---|---|
Temperature (Metric) Range | 0°C to 80°C |
Tip Type | Standard |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Connector Types | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Connector Types | Price | Quantity & Availability | |||
11798452
|
Fisherbrand™
|
25 mm | 5 mm | BNC |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11788462
|
Fisherbrand™
|
75 mm | 12 mm | DIN |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Reinforced
- Excellent for meat, cheese, fish, fruit
- Electrolyte KCI 3M + AgCl
- pH range of 0 to 12
- Temperature range of 0° to 80°C
Specifications
0 to 12 | |
0°C to 80°C | |
Standard |