Learn More
Testo™ Type T Super-Quick Needle Probe
For measurements without visible penetration hole.
Brand:  Testo™ 0628 0027
Product Code. 10731013
  • 945.00 SEK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
  • Ideal for hamburgers, steaks, etc.
  • Waterproof
  • Reaction time of 2 seconds
  • Measuring range of −50° to +250°C
Specifications
Specifications
1.4 mm
Reaction time of 2 seconds
-60°C
±0.2°C from −20° to +70°C
150 mm
250°C
-50°C to +250°C
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Testo™ Type T Super-Quick Needle Probe > Type T Super-Quick Needle Probe