Learn More
PEPD Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12218-1-AP-150UL
 View more versions of this product
Product Code. 16889793
  • 4652.90 SEK / 150µL
Description
Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids.Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids.
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PEPD Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated