missing translation for 'onlineSavingsMsg'
Learn More
Learn More
peptide YY Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24294-1-AP-20UL
This item is not returnable.
View return policy
Description
Peptide tyrosine-tyrosine amide (3-36), or PYY (3-36), is a major metabolite of the gut hormone PYY and is produced by the action of dipeptidyl peptidase IV on PYY in the intestinal brush border and in the circulation. PYY (3-36) is a potent inhibitor of food intake in rats and humans, acting selectively on the Y2 receptor to inhibit orexigenic NPY neurons in the arcuate nucleus, thus disinhibiting anorexigenic POMC neurons. The anorexigenic effect of PYY (3-36) is additive to that of the co-secreted gut hormone GLP-1.Specifications
peptide YY | |
Polyclonal | |
Unconjugated | |
PYY | |
Peptide tyrosine tyrosine, peptide YY, PYY, PYY I, PYY II, PYY1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5697 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10082 | |
PYY | |
peptide YY Fusion Protein Ag17939 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
peptide YY Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction