Learn More
Honeywell™ PERFECT POLY™ PU First Gray Gloves
Honeywell™ PERFECT POLY™ PU First Gray Gloves for handling of fine objects in a dry environment, or slightly greasy and dirty objects.
23.87 SEK - 70.00 SEK
Specifications
Certifications/Compliance | EN 420-2003 Basic standard, EN 388-2016 Mechanical risk |
---|---|
Description | Knitted Coating, 13 gauge |
Coating Material | Polyurethane |
Color | Gray |
Material | Polyamide |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12675293
|
Honeywell™
2100250-07 |
7 |
70.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
12685293
|
Honeywell™
2100250-08 |
8 |
70.00 SEK
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12695293
|
Honeywell™
2100250-09 |
9 |
46.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
15826685
|
Honeywell™
2100250-06 |
6 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15856685
|
Honeywell™
2100250-09 |
9 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15846685
|
Honeywell™
2100250-08 |
8 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Gray polyamide knitted glove
- Gray polyurethane coating at the palm and fingertips
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back and PU coating optimise the glove's breathability
- Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
- The PU coating ensures a good grip
- Non pilling, the polyamide is suitable for precision work
- The gloves are certified as silicone-free to ensure their use in painting environment
- Gray color makes it more soilproof, thus ensuring a longer life
Specifications
EN 420-2003 Basic standard, EN 388-2016 Mechanical risk | |
Polyurethane | |
Polyamide |
Knitted Coating, 13 gauge | |
Gray | |
25 cm |