Learn More
Honeywell Research Chemicals 2.5LT Petroleum special, ~18% aromatics basis, bp 180-220 oC
Brand:  Honeywell Research Chemicals 77370-2.5L
2.5LT Petroleum special, ~18% aromatics basis, bp 180-220 oC
Additional Details : CAS Number : 64742-82-1 Weight : 1.95000kg
Product Code. 15697060