Learn More
PGAM1 Mouse anti-Human, Unconjugated, Clone: 2H2A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67470-1-IG-20UL
Description
The PGAM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PGAM1. This antibody recognizes human, mouse, rat antigen. The PGAM1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PGAM1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
2H2A9 | |
Unconjugated | |
20 μL | |
5223 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.