missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGP9.5 / UchL1 (pan-Neuronal Marker) Mouse, Clone: rUCHL1/775, enQuireBio™
Mouse Monoclonal Antibody
2035.00 SEK - 5965.00 SEK
Specifications
Antigen | PGP9.5 / UchL1 (pan-Neuronal Marker) |
---|---|
Clone | rUCHL1/775 |
Concentration | 200ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15981048
|
EnquireBio
7345-MSM5-P0 |
20 ÎĽg | N/A | ||||||
15991048
|
EnquireBio
7345-MSM5-P1 |
100 ÎĽg |
5965.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
PGP9.5 / UchL1 (pan-Neuronal Marker) | |
200ÎĽg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human UCHL1 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
20-30kDa |
rUCHL1/775 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
7345 | |
IgG1 Îş | |
Protein A/G purified from Bioreactor concentrate. | |
Cytoplasmic. Endoplasmic Reticulum membrane | |
Cerebellum |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PGP9.5 / UchL1 (pan-Neuronal Marker) Mouse, Clone: rUCHL1/775, enQuireBio™