Learn More
pH 9.00 Buffer Solution at 25°C, Reagecon Product Code.: 11454559
pH 9.00 Buffer Solution at 25°C, Reagecon
BIOPHARMA
Product Code. 11454559 Shop All Reagecon Products
1 L, 1 L
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
Product Code. 11454559
Brand: Reagecon 109025
Tested, certified, NIST traceable, and shelf-stable buffer solution.
  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Description Accuracy ± 0.01 pH units
pH 9.00 ±0.01
Physical Form Liquid
Quantity 1 L
Packaging HDPE Bottle GL45
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number192525E1 Certificate TypeCertificate of Analysis Date25/05/2025 Product Code109025
Lot Number192525A1 Certificate TypeCertificate of Analysis Date25/02/2025 Product Code109025
Lot Number192524G1 Certificate TypeCertificate of Analysis Date19/11/2024 Product Code109025
Lot Number192524B1 Certificate TypeCertificate of Analysis Date29/05/2024 Product Code109025
Lot Number192524B1 (1) Certificate TypeCertificate of Analysis Date15/03/2024 Product Code109025
Product Content Correction
Product Title
pH 9.00 Buffer Solution at 25°C, Reagecon >