Learn More
PHACS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11491-1-AP-150UL
 View more versions of this product
Product Code. 16888603
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Does not catalyze the synthesis of 1-aminocyclopropane- 1-carboxylate but is capable of catalyzing the deamination of L- vinylglycine.
Specifications
Specifications
PHACS
Polyclonal
Unconjugated
ACCS
ACC synthase like protein 1, ACCS, ACS, PHACS
Rabbit
Antigen Affinity Chromatography
RUO
311218, 329470, 84680
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2AIG8, Q96QU6
Accs
PHACS Fusion Protein Ag2056
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PHACS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated