missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHACTR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PHACTR4 |
---|---|
Concentration | 0.34 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881604
|
Proteintech
13408-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871604
|
Proteintech
13408-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the phosphatase and actin regulator (PHACTR) family. Other PHACTR family members have been shown to inhibit protein phosphatase 1 (PP1) activity, and the homolog of this gene in the mouse has been shown to interact with actin and PP1. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
PHACTR4 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q501J7, Q8IZ21 | |
100169, 65979, 682920 | |
PHACTR4 Fusion Protein Ag4054 | |
Primary | |
-20°C | |
PHACTR4 |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PHACTR4 | |
PHACTR4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PHACTR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech