missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24279-1-AP-150UL
This item is not returnable.
View return policy
Description
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.Specifications
PHKA1 | |
Polyclonal | |
Unconjugated | |
PHKA1 | |
PHKA, PHKA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18679, 5255, 64561 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18826, P46020, Q64649 | |
Phka1 | |
PHKA1 Fusion Protein Ag17998 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PHKA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction