missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24658-1-AP-20UL
This item is not returnable.
View return policy
Description
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.Specifications
PHKA2 | |
Polyclonal | |
Unconjugated | |
PHKA2 | |
GSD9A, PHK, PHKA2, PHKLA, PYK, PYKL, XLG, XLG2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110094, 5256, 678739 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P46019, Q8BWJ3 | |
PHKA2 | |
PHKA2 Fusion Protein Ag9473 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PHKA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction