Learn More
Phosphate Buffer Solution, pH 7, Honeywell™ Fluka™
Brand: Honeywell Fluka 73173-10L
Quantity | 10 L |
---|---|
Packaging | Poly pail |
This item is not returnable.
View return policy
Description
Specifications
10 L | |
Poly pail |
Product Content Correction
Product Title
Phosphate Buffer Solution, pH 7, Honeywell™ Fluka™ > 10L; Poly pail
Spot an opportunity for improvement?Share a Content Correction