missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Actin alpha/gamma (Tyr53, Tyr55) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538402
This item is not returnable.
View return policy
Description
Actins are highly conserved proteins that are involved in various types of cell motility, and maintece of the cytoskeleton. In vertebrates, three main groups of actin isoforms, alpha, beta and gamma have been identified. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exist in most cell types as components of the cytoskeleton, and as mediators of internal cell motility. Actin gamma 1 is a cytoplasmic actin found in nonmuscle cells.
Specifications
Phospho-Actin alpha/gamma (Tyr53, Tyr55) | |
Polyclonal | |
Unconjugated | |
ACTG1 | |
ACT, ACTG, BRWS2, DFNA20, DFNA26, actin, cytoplasmic 2, cytoskeletal gamma-actin, gamma-actin, actin gamma, cytoplasmic actin, ACTG1, Atcb | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11461, 11465, 60, 71 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P60709, P63261, P60710, P63260 | |
ACTB, ACTG1 | |
A synthetic phosphopeptide derived from human Actin-pan (alpha/gamma) around the phosphorylation site of Tyr53/55 (D-S-YP-V-G) | |
100 ÎĽg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phospho-Actin alpha/gamma (Tyr53, Tyr55) Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction