missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-C/EBP beta (Thr235, Thr188) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104823
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of CEBP Beta only when phosphorylated at Thr235 in human, or Thr188 in mouse and rat.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.
Specifications
Phospho-C/EBP beta (Thr235, Thr188) | |
Polyclonal | |
Unconjugated | |
Cebpb | |
AGP/EBP; C EBP; c/EBP beta; C/EBPb; C/EBPbeta; C/EBP-beta; C/EBP-related protein 2; CCAAT enhancer binding protein beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer binding protein beta; CCAAT/enhancer-binding protein beta; CEBPB; Crp2; Il6dbp; IL-6DBP; interleukin 6-dependent DNA-binding protein; interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nfil6; Nf-il6; NF-M; nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; PP9092; Sfb; SF-B; Silencer factor B; TCF5; TCF-5; transcription factor 5; transcription factor C/EBP beta | |
Rabbit | |
sequential chromatography | |
RUO | |
1051, 12608, 24253 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P17676, P21272, P28033 | |
Cebpb | |
A synthesized peptide derived from human CEBPB(Accession P17676), corresponding to amino acid residues around phosphorylated Thr235. | |
100 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Phospho-C/EBP beta (Thr235, Thr188) Polyclonal Antibody > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction