Learn More
Invitrogen™ phospho-Tyrosine Monoclonal Antibody (pY20), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 46-5001-42
Description
The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
Specifications
phospho-Tyrosine | |
Monoclonal | |
5 ÎĽL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 Tests | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid | |
Chemical |
Flow Cytometry | |
pY20 | |
PerCP-eFluor 710 | |
Phosphotyrosine; pTyr; pY | |
Affinity chromatography | |
RUO | |
Chemical | |
Antibody | |
IgG2b Îş |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.