missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20421-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
PIGM encodes a transmembrane protein that is located in the endoplasmic reticulum and is involved in GPI-anchor biosynthesis. The glycosylphosphatidylinositol (GPI)-anchor is a glycolipid which contains three mannose molecules in its core backbone. The GPI-anchor is found on many blood cells and serves to anchor proteins to the cell surface. PIGM encodes a mannosyltransferase, GPI-MT-I, that transfers the first mannose to GPI on the lumenal side of the endoplasmic reticulum.Specifications
PIGM | |
Polyclonal | |
Unconjugated | |
PIGM | |
GPI mannosyltransferase 1, GPI mannosyltransferase I, GPI MT I, PIG M, PIGM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67556, 79112, 93183 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8C2R7, Q9EQY6, Q9H3S5 | |
PIGM | |
PIGM Fusion Protein Ag14245 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |