Learn More
PIR Mouse anti-Human, Unconjugated, Clone: 2A10E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67516-1-IG-150UL
 View more versions of this product
Product Code. 16890186
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The PIR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PIR. This antibody recognizes human, mouse, rat antigen. The PIR antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PIR
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
2A10E4
Unconjugated
150 μL
8544
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PIR Mouse anti-Human, Unconjugated, Clone: 2A10E4, Proteintech > 150 μL