Learn More
PKC epsilon Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20877-1-AP-150UL
 View more versions of this product
Product Code. 16880455
  • 4652.90 SEK / 150µL
Description
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PKC epsilon Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated