Learn More
PKM2-Specific Mouse anti-Human, CoraLite 594, Clone: 1C11C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60268-100UL
Description
The PKM2-specific antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human PKM2-specific. This antibody recognizes human, mouse, rat antigen. The PKM2-specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
PKM2-Specific | |
Monoclonal | |
CoraLite 594 | |
100 ÎĽL | |
5315 | |
IgG1 |
Immunofluorescence | |
1C11C7 | |
Mouse | |
Primary | |
Human, Rat, Mouse |