missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKMYT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55098-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to PKMYT1.
This gene encodes a protein serine/threonine kinase that is activated when bound to the GTP-bound form of Rho. The small GTPase Rho regulates formation of focal adhesions and stress fibers of fibroblasts, as well as adhesion and aggregation of platelets and lymphocytes by shuttling between the inactive GDP-bound form and the active GTP-bound form. Rho is also essential in cytokinesis and plays a role in transcriptional activation by serum response factor. This protein, a downstream effector of Rho, phosphorylates and activates LIM kinase, which in turn, phosphorylates cofilin, inhibiting its actin-depolymerizing activity.Specifications
PKMYT1 | |
Polyclonal | |
Unconjugated | |
PKMYT1 | |
MYT1, Myt1 kinase, PKMYT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
268930, 287101, 9088 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99640, Q9ESG9 | |
Pkmyt1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PKMYT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction