missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PLA2G1B (Human) Recombinant Protein
Human PLA2G1B full-length ORF ( AAH05386, 17 a.a. - 70 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005319-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: SGISPRAVWQFRKMIKCVIPGSDPFLEYNNYGCYCGLGGSGTPVDELDKQKQRV
Specifications
AAH05386 | |
phospholipase A2, group IB (pancreas) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PLA2G1B | |
GST |
5319 | |
Wheat germ expression system | |
10 μg | |
MGC119834, MGC119835, PLA2, PLA2A, PPLA2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |