missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4F Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21411-1-AP-150UL
This item is not returnable.
View return policy
Description
Calcium-dependent phospholipase A2 that selectively hydrolyzes glycerophospholipids in the sn-2 position. Has higher enzyme activity for phosphatidylethanolamine than phosphatidylcholine.Specifications
PLA2G4F | |
Polyclonal | |
Unconjugated | |
PLA2G4F | |
cPLA2 zeta, Phospholipase A2 group IVF, phospholipase A2, group IVF, PLA2G4F, PLA2G4F/Z | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
255189 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68DD2 | |
PLA2G4F | |
PLA2G4F Fusion Protein Ag15571 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PLA2G4F Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction