Learn More
Cowie Technology™ Cross-Shape Stirring Magnet, PTFE
A very stable general purpose stirrer. The COWIE TECHNOLOGY™ Cross-Shape Stirring Magnet has a similar action to a cylindrical stirring magnet, but gives more turbulence at low speed.
Brand:  Cowie Technology™ 001.2407
Product Code. 10620283
  • 117.00 SEK / Each
Only null left
Estimated Shipment: 16-07-2025
Add to basket
Description
Description
  • Stirring magnet for easy and convenient mixing
  • PTFE construction
Specifications
Specifications
Stirring Magnet
3.375 in.
60 mm
PTFE
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cowie Technology™ Cross-Shape Stirring Magnet, PTFE > 40mm x 7mm