Learn More
Fisherbrand™ Plastic PP Syringes, Luer Slip Product Code.: 12951031
Fisherbrand™ Plastic PP Syringes, Luer Slip
Product Code. p-7113856 Shop All Fisherbrand Products
20 mL, Pack of 100
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
20 mL
Unit Size:
Pack of 100
Product Code. 12951031
Edge
HPLC certified, non sterile disposable syringes made of solvent robust polypropylene
  • Plastic Disposable Syringe
  • Polypropylene
  • Luer Slip
  • Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
  • No silicone or oil lubricant is required in the barrel
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Volume (Metric) 20 mL
Connection Type Leur Slip
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24157
    Expires2025-08-31
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/07/2025 and 31/08/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Fisherbrand™ Plastic PP Syringes, Luer Slip >