Learn More
PLCG1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28362-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The PLCG1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PLCG1. This antibody recognizes human antigen. The PLCG1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PLCG1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
5335 | |
IgG |
ELISA, Western Blot | |
400 μg/mL | |
Rabbit | |
Primary | |
Human |