Learn More
PLCG1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28362-1-AP-20UL
 View more versions of this product
Product Code. 16886105
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The PLCG1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PLCG1. This antibody recognizes human antigen. The PLCG1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PLCG1
Polyclonal
Unconjugated
20 μL
5335
IgG
Western Blot, ELISA
400 μg/mL
Rabbit
Primary
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PLCG1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL