Learn More
PLCG2 Mouse anti-Human, Unconjugated, Clone: 2F8H5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67011-1-IG-20UL
 View more versions of this product
Product Code. 16889375
  • 1645.54 SEK / 20µL
Description
The PLCG2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLCG2. This antibody recognizes human antigen. The PLCG2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PLCG2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
2F8H5
Unconjugated
20 μL
5336
IgG1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PLCG2 Mouse anti-Human, Unconjugated, Clone: 2F8H5, Proteintech > 20 μL