Learn More
PLCG2 Mouse anti-Human, Unconjugated, Clone: 2F8H5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67011-1-IG-20UL
Beschreibung
The PLCG2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLCG2. This antibody recognizes human antigen. The PLCG2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
PLCG2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
2F8H5 | |
Unconjugated | |
20 μL | |
5336 | |
IgG1 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.