missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLOD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21214-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to PLOD2.
The protein encoded by this gene is a membrane-bound homodimeric enzyme that is localized to the cisternae of the rough endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VIB have deficiencies in lysyl hydroxylase activity. Mutations in the coding region of this gene are associated with Bruck syndrome. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
PLOD2 | |
Polyclonal | |
Unconjugated | |
PLOD2 | |
LH2, Lysyl hydroxylase 2, PLOD2-Specific, TLH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26432, 300901, 5352 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00469, Q811A3, Q9R0B9 | |
PLOD2 | |
PLOD2-Specific Fusion Protein Ag15614 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PLOD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction