missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLRG1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PLRG1 |
---|---|
Concentration | 0.28 mg/mL |
Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899293
|
Proteintech
11914-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889293
|
Proteintech
11914-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plrg1 is a protein coding gene involved in pre-mRNA splicing as component of the spliceosome. Component of the PRP19-CDC5L complex that forms an integral part of the spliceosome and is required for activating pre-mRNA splicing.Specifications
PLRG1 | |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
O43660 | |
5356 | |
PLRG1 Fusion Protein Ag2527 | |
Primary | |
-20°C | |
PLRG1 |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Pleiotropic regulator 1, PLRG1, PRL1 | |
PLRG1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PLRG1 Rabbit anti-Human, Polyclonal, Proteintech