missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PLXNA2 (Human) Recombinant Protein
Human PLXNA2 full-length ORF ( AAH32125, 1 a.a. - 164 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005362-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MHSQVFSAFPYSLPLRFWVNVIKNPQFVFDIHKGSITDACLSVVAQTFMDSCSTSEHRLDKDSPSNKLLYAKDIPSYKSWVERYYADIAKLPAISDQDMNAYLAEQSRLHAVEFNMLSALNEIYSYVSKYSEELIGALEQDEQARRQRLAYKVEQLINAMSIES
Specifications
AAH32125 | |
plexin A2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PLXNA2 | |
GST |
5362 | |
Wheat germ expression system | |
10 μg | |
FLJ11751, FLJ30634, KIAA0463, OCT, PLXN2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |