missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMAL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16965-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
PNMAL1 | |
Polyclonal | |
Unconjugated | |
PNMA8A | |
PNMA like 1, PNMA like protein 1, PNMAL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55228 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86V59 | |
PNMA8A | |
PNMAL1 Fusion Protein Ag10698 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PNMAL1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction