Learn More
Podoplanin Mouse anti-Human, Unconjugated, Clone: 1D9F3, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Podoplanin |
---|---|
Clone | 1D9F3 |
Concentration | 1000 ÎĽg/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800066
|
Proteintech
67432-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890056
|
Proteintech
67432-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Podoplanin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Podoplanin. This antibody recognizes human antigen. The Podoplanin antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Podoplanin | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
10630 | |
Primary |
1D9F3 | |
Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.