missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12254-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a component of the RNA polymerase I and RNA polymerase III complexes, which function in the synthesis of ribosomal RNA precursors and small RNAs, respectively. Mutations in this gene are a cause of Treacher Collins syndrome (TCS), a craniofacial development disorder. Alternative splicing results in multiple transcript variants.Specifications
POLR1D | |
Polyclonal | |
Unconjugated | |
POLR1D | |
AC19, hRPA19, POLR1C, POLR1D, RPA16, RPA9, RPAC2, RPC16, RPO1 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20018, 360762, 51082 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97304, Q9Y2S0 | |
Polr1d | |
POLR1D Fusion Protein Ag2904 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
POLR1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction