Learn More
Scientific Laboratory Supplies™ SLS Select Drying Rack
Polyethylene-coated draining/drying rack for laboratory glassware. Wall mounted.
Brand:  Scientific Laboratory Supplies™ DR340
Product Code. 12766045
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Wall mounting
  • Polyethylene-coated rack, with pegs
  • Angled brackets support the rack about 25mm away from the wall surface
Specifications
Specifications
Draining Rack
1 Each
560 mm
Polyethylene-coated Metal Frame
560 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Scientific Laboratory Supplies™ SLS Select Drying Rack > 510 x 480mm